Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Supplier:  Analytical Sales & Services LUM22418LS660
Encompass
LumidoxII 24 Well, 18mm Spaced 660-Deep Red LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05243