Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Catalog No. 5000805246
Encompass
LumidoxII 24 Well, 18mm Spaced 660-Deep Red LED Array, with Lens Mat and Active Cooling Base.
Catalog No. 50-008-05246 Supplier Analytical Sales & Services Supplier No. LUM22418LA660
LumidoxII 24 Well, 18mm Spaced 660-Deep Red LED Array, with Lens Mat and Active Cooling Base.