Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Catalog No. 5000805569
Encompass
LumidoxII 24 Well, 18mm Spaced 527-GREEN LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05569 Supplier Analytical Sales & Services Supplier No. LUM22418LS527
LumidoxII 24 Well, 18mm Spaced 527-GREEN LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)