Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Catalog No. 5000806375
Encompass
LumidoxII 24 Well, 18mm Spaced UV385 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06375 Supplier Analytical Sales & Services Supplier No. LUM22418LS385
LumidoxII 24 Well, 18mm Spaced UV385 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)