Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Catalog No. 5000806815
Encompass
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06815 Supplier Analytical Sales & Services Supplier No. LUM22418LS405
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)