Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Supplier: Analytical Sales & Services LUM22418LS810
LumidoxII 24 Well, 18mm Spaced 810-IR LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)