Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Catalog No. 5000807227
Encompass
LumidoxII 24 Well, 18mm Spaced 420-VIOLET LED Array, with Lens Mat and Flow-through Base (requires external chiller)
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.
Catalog No. 50-008-07227 Supplier Analytical Sales & Services Supplier No. LUM22418LF420
LumidoxII 24 Well, 18mm Spaced 420-VIOLET LED Array, with Lens Mat and Flow-through Base (requires external chiller)
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.