Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays
Supplier: Analytical Sales & Services LUM22418LF445
LumidoxII 24 Well, 18mm Spaced 445-INDIGO LED Array, with Lens Mat and Flow-through Base (requires external chiller)
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.