Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays for Cell/Bio Work - 527-Green
Catalog No. 5000805410
Encompass
LumidoxII 24 Well, 18mm Spaced 527-Green LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05410 Supplier Analytical Sales & Services Supplier No. BIOLUM22418LA527
LumidoxII 24 Well, 18mm Spaced 527-Green LED Array, with Lens Mat and Active Cooling Base