Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays for Cell/Bio Work - UV365
Catalog No. 5000806433
Encompass
LumidoxII 24 Well, 18mm Spaced UV365 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-06433 Supplier Analytical Sales & Services Supplier No. BIOLUM22418LA365
LumidoxII 24 Well, 18mm Spaced UV365 LED Array, with Lens Mat and Active Cooling Base