Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays for Cell/Bio Work - UV405
Catalog No. 5000807161
Encompass
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-07161 Supplier Analytical Sales & Services Supplier No. BIOLUM22418LA405
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Active Cooling Base