Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays (with Lens Mat and Solid Base, for 1 Dram TCR) - 420-Violet
Catalog No. 5000805794
Encompass
LumidoxII 24 Well, 18mm Spaced 420-Violet LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05794 Supplier Analytical Sales & Services Supplier No. TCRLUM22418LS420
LumidoxII 24 Well, 18mm Spaced 420-Violet LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)