Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays (with Lens Mat and Solid Base, for 1 Dram TCR) - 470-Blue
Supplier:  Analytical Sales & Services TCRLUM22418LS470
Encompass
LumidoxII 24 Well, 18mm Spaced 470-Blue LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-04940