Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays (with Lens Mat and Solid Base, for 1 Dram TCR) - 527-Green
Catalog No. 5000805797
Encompass
LumidoxII 24 Well, 18mm Spaced 527-Green LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05797 Supplier Analytical Sales & Services Supplier No. TCRLUM22418LS527
LumidoxII 24 Well, 18mm Spaced 527-Green LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)