Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays (with Lens Mat and Solid Base, for 1 Dram TCR) - UV395
Catalog No. 5000805792
Encompass
LumidoxII 24 Well, 18mm Spaced UV395 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05792 Supplier Analytical Sales & Services Supplier No. TCRLUM22418LS395
Add to Cart
Add to Cart
LumidoxII 24 Well, 18mm Spaced UV395 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)