Learn More
Analytical Sales & Services Lumidox II 24-Well, 18mm Spacing LED Arrays (with Lens Mat and Solid Base, for 1 Dram TCR) - UV405
Catalog No. 5000805793
Encompass
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05793 Supplier Analytical Sales & Services Supplier No. TCRLUM22418LS405
LumidoxII 24 Well, 18mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)