Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Catalog No. 5000806374
Encompass
LumidoxII 24 Well, 9mm Spaced UV395 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06374 Supplier Analytical Sales & Services Supplier No. LUM224LS395
LumidoxII 24 Well, 9mm Spaced UV395 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)