Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Supplier:  Analytical Sales & Services LUM224LS375
Encompass
LumidoxII 24 Well, 9mm Spaced UV375 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06814