Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Catalog No. 5000806817
Encompass
LumidoxII 24 Well, 9mm Spaced 445-INDIGO LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06817 Supplier Analytical Sales & Services Supplier No. LUM224LS445
Add to Cart
Add to Cart
LumidoxII 24 Well, 9mm Spaced 445-INDIGO LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)