Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Catalog No. 5000807190
Encompass
LumidoxII 24 Well, 9mm Spaced UV385 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-07190 Supplier Analytical Sales & Services Supplier No. LUM224LS385
LumidoxII 24 Well, 9mm Spaced UV385 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)