Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Catalog No. 5000805215
Encompass
LumidoxII 24 Well, 9mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05215 Supplier Analytical Sales & Services Supplier No. LUM224LS405
LumidoxII 24 Well, 9mm Spaced UV405 LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)