Learn More
Analytical Sales & Services Lumidox II 24-Well, 9mm Spaced LED Arrays
Catalog No. 5000805156
Encompass
LumidoxII 24 Well, 9mm Spaced 630-RED LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05156 Supplier Analytical Sales & Services Supplier No. LUM224LA630
LumidoxII 24 Well, 9mm Spaced 630-RED LED Array, with Lens Mat and Active Cooling Base