Learn More
Analytical Sales & Services Lumidox II 96-Position Discovery LED Array 2 (UV405, Indigo, Blue & Amber) - Flow-Through Base
Catalog No. 5000805433
Encompass
LumidoxII 96 Well Discovery LED Array 2, with Lens Mat and Flow-through Base (requires external chiller)\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.
Catalog No. 50-008-05433 Supplier Analytical Sales & Services Supplier No. LUM296LFG2
LumidoxII 96 Well Discovery LED Array 2, with Lens Mat and Flow-through Base (requires external chiller)\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.