Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805524
Encompass
LumidoxII 96 Well, 660-DEEP RED LED Array, with Lens Mat and Flow-through Base. For deep red, near IR photocatalysis (requires external chiller )\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.
Catalog No. 50-008-05524 Supplier Analytical Sales & Services Supplier No. LUM296LF660
Add to Cart
Add to Cart
LumidoxII 96 Well, 660-DEEP RED LED Array, with Lens Mat and Flow-through Base. For deep red, near IR photocatalysis (requires external chiller )\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.