Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805574
Encompass
LumidoxII 96 Well, 590-AMBER LED Array, with Diffuse Mat and Active Cooling Base
Catalog No. 50-008-05574 Supplier Analytical Sales & Services Supplier No. LUM296DA590
LumidoxII 96 Well, 590-AMBER LED Array, with Diffuse Mat and Active Cooling Base