Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000806383
Encompass
LumidoxII 96 Well, UV375 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06383 Supplier Analytical Sales & Services Supplier No. LUM296DS375
LumidoxII 96 Well, UV375 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)