Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000806609
Encompass
LumidoxII 96 Well, UV395 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06609 Supplier Analytical Sales & Services Supplier No. LUM296DS395
Add to Cart
Add to Cart
LumidoxII 96 Well, UV395 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)