Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Supplier:  Analytical Sales & Services LUM296DS810
Encompass
LumidoxII 96 Well, 810-IR LED Array, with Diffuse Mat and Solid Base. (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06611