Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000806695
Encompass
LumidoxII 96 Well, 810-IR LED Array, with Lens Mat and Solid Base. (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06695 Supplier Analytical Sales & Services Supplier No. LUM296LS810
LumidoxII 96 Well, 810-IR LED Array, with Lens Mat and Solid Base. (requires Thermal Transfer Deck and external chiller)