Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000806774
Encompass
LumidoxII 96 Well, 590-AMBER LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06774 Supplier Analytical Sales & Services Supplier No. LUM296LS590
LumidoxII 96 Well, 590-AMBER LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)