Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805165
Encompass
LumidoxII 96 Well, 470-BLUE LED Array, with Diffuse Mat and Flow-through Base (requires external chiller )\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.
Catalog No. 50-008-05165 Supplier Analytical Sales & Services Supplier No. LUM296DF470
LumidoxII 96 Well, 470-BLUE LED Array, with Diffuse Mat and Flow-through Base (requires external chiller )\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.