Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Supplier:  Analytical Sales & Services LUM296DS730
Encompass
LumidoxII 96 Well, 730-IR LED Array, with Diffuse Mat and Solid Base. (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05167