Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Supplier:  Analytical Sales & Services LUM296LSWHT
Encompass
LumidoxII 96 Well, WHITE LED Array, with Lens Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-06856