Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Supplier:  Analytical Sales & Services LUM296DA375
Encompass
LumidoxII 96 Well, UV375 LED Array, with Diffuse Mat and Active Cooling Base
Catalog No. 50-008-07281