Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Supplier: Analytical Sales & Services LUM296LF405
LumidoxII 96 Well, UV405 LED Array, with Lens Mat and Flow-through Base (requires external chiller )\n
Note: Flow-through base LED arrays can only be used with an independent, dedicated recirculating chiller circuit. They cannot be daisy chained with any other hardware.