Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805191
Encompass
LumidoxII 96 Well, UV405 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05191 Supplier Analytical Sales & Services Supplier No. LUM296DS405
LumidoxII 96 Well, UV405 LED Array, with Diffuse Mat and Solid Base (requires Thermal Transfer Deck and external chiller)