Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805230
Encompass
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Active Cooling Base. For deep red, near IR photocatalysis.
Catalog No. 50-008-05230 Supplier Analytical Sales & Services Supplier No. LUM296DA660
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Active Cooling Base. For deep red, near IR photocatalysis.