Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays
Catalog No. 5000805256
Encompass
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Solid Base. For deep red, near IR photocatalysis (requires Thermal Transfer Deck and external chiller)
Catalog No. 50-008-05256 Supplier Analytical Sales & Services Supplier No. LUM296DS660
LumidoxII 96 Well, 660-DEEP RED LED Array, with Diffuse Mat and Solid Base. For deep red, near IR photocatalysis (requires Thermal Transfer Deck and external chiller)