Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - 420-Violet
Catalog No. 5000805934
Encompass
LumiDox Gen2, 96 Well, 420-VIOLET LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05934 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA420
LumiDox Gen2, 96 Well, 420-VIOLET LED Array, with Lens Mat and Active Cooling Base