Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - 527-Green
Catalog No. 5000806831
Encompass
LumiDox Gen2, 96 Well, 527-GREEN LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-06831 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA527
LumiDox Gen2, 96 Well, 527-GREEN LED Array, with Lens Mat and Active Cooling Base