Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - 590-Amber
Catalog No. 5000807177
Encompass
LumiDox Gen2, 96 Well, 590-AMBER LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-07177 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA590
LumiDox Gen2, 96 Well, 590-AMBER LED Array, with Lens Mat and Active Cooling Base