Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - 630-Red
Catalog No. 5000805414
Encompass
LumiDox Gen2, 96 Well, 630-RED LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05414 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA630
LumiDox Gen2, 96 Well, 630-RED LED Array, with Lens Mat and Active Cooling Base