Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - 660-Deep Red
Catalog No. 5000806832
Encompass
LumiDox Gen2, 96 Well, 660-DEEP RED LED Array, with Lens Mat and Active Cooling Base. For deep red, near IR photocatalysis.
Catalog No. 50-008-06832 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA660
Add to Cart
Add to Cart
LumiDox Gen2, 96 Well, 660-DEEP RED LED Array, with Lens Mat and Active Cooling Base. For deep red, near IR photocatalysis.