Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - UV375
Catalog No. 5000805411
Encompass
LumiDox Gen2, 96 Well, UV375 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05411 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA375
LumiDox Gen2, 96 Well, UV375 LED Array, with Lens Mat and Active Cooling Base