Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays for Cell/Bio Work - UV385
Catalog No. 5000805412
Encompass
LumiDox Gen2, 96 Well, UV385 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05412 Supplier Analytical Sales & Services Supplier No. BIOLUM296LA385
LumiDox Gen2, 96 Well, UV385 LED Array, with Lens Mat and Active Cooling Base