Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Catalog No. 5000805344
Encompass
LumiDox Gen2, 96 Well, UV365 LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05344 Supplier Analytical Sales & Services Supplier No. LUM296LA3651
Add to Cart
Add to Cart
LumiDox Gen2, 96 Well, UV365 LED Array, with Lens Mat and Active Cooling Base