Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Supplier: Analytical Sales & Services LUM296LS6601
LumiDox Gen2, 96 Well, 660-DEEP RED LED Array, with Lens Mat and Solid Base. For deep red, near IR photocatalysis.