Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Catalog No. 5000805471
Encompass
LumiDox Gen2, 96 Well, 445-INDIGO LED Array, with Lens Mat and Active Cooling Base
Catalog No. 50-008-05471 Supplier Analytical Sales & Services Supplier No. LUM296LA4451
LumiDox Gen2, 96 Well, 445-INDIGO LED Array, with Lens Mat and Active Cooling Base