Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Catalog No. 5000805517
Encompass
LumiDox Gen2, 96 Well, 730-IR LED Array, with Lens Mat and Solid Base. For deep red, near IR photocatalysis.
Catalog No. 50-008-05517 Supplier Analytical Sales & Services Supplier No. LUM296LS7301
LumiDox Gen2, 96 Well, 730-IR LED Array, with Lens Mat and Solid Base. For deep red, near IR photocatalysis.