Learn More
Analytical Sales & Services Lumidox II 96-Well LED Arrays (Copy)
Catalog No. 5000807240
Encompass
LumiDox Gen2, 96 Well, 445-INDIGO LED Array, with Diffuse Mat and Active Cooling Base
Catalog No. 50-008-07240 Supplier Analytical Sales & Services Supplier No. LUM296DA4451
Add to Cart
Add to Cart
LumiDox Gen2, 96 Well, 445-INDIGO LED Array, with Diffuse Mat and Active Cooling Base